Order Entry
Netherlands
ContactUsLinkComponent
1-(2-Chloro-4-fluorobenzyl)-1H-pyrazol-3-amine
1-(2-Chloro-4-fluorobenzyl)-1H-pyrazol-3-amine
Catalog # APOSPC410296-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(2-Chloro-4-fluorobenzyl)-1H-pyrazol-3-amine
Catalog # APOSPC410296-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :225.65 g/mol
Storage Temperature:Ambient
CAS Number:1001757-50-1
Formula:C₁₀H₉ClFN₃
MDL Number:MFCD02254006