4-Chloro-1-methyl-5-(trifluoromethyl)-1H-pyrazole-3-carboxylic acid
Catalog # APOSPC410290-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:228.556 g/mol
Storage Temperature:Ambient
CAS Number:1001519-38-5
Formula:C₆H₄ClF₃N₂O₂
MDL Number:MFCD03419816