1-Ethyl-5-(trifluoromethyl)-1H-pyrazole-3-carbohydrazide
Catalog # APOSPC410286-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:222.168 g/mol
Storage Temperature:Ambient
CAS Number:1001519-13-6
Formula:C₇H₉F₃N₄O
MDL Number:MFCD05667186