Order Entry
Netherlands
ContactUsLinkComponent
3-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide
3-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide
Catalog # APOSPC410272-1G
Supplier:  Apollo Scientific
CAS Number:  
3-[5-Methyl-3-(trifluoromethyl)-1H-pyrazol-1-yl]propanehydrazide
Catalog # APOSPC410272-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :236.194 g/mol
Storage Temperature:Ambient
CAS Number:1001518-86-0
Formula:C₈H₁₁F₃N₄O
MDL Number:MFCD03419696