Order Entry
Netherlands
ContactUsLinkComponent
2-[3-(Difluoromethyl)-5-methyl-1H-pyrazol-1-yl]acetohydrazide
2-[3-(Difluoromethyl)-5-methyl-1H-pyrazol-1-yl]acetohydrazide
Catalog # APOSPC410268-1G
Supplier:  Apollo Scientific
CAS Number:  
2-[3-(Difluoromethyl)-5-methyl-1H-pyrazol-1-yl]acetohydrazide
Catalog # APOSPC410268-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :204.177 g/mol
Storage Temperature:Ambient
CAS Number:1001518-82-6
Formula:C₇H₁₀F₂N₄O
MDL Number:MFCD03419679