Order Entry
Netherlands
ContactUsLinkComponent
4-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazole-5-carbohydrazide
4-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazole-5-carbohydrazide
Catalogus # APOSPC410265-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Chloro-1-methyl-3-(trifluoromethyl)-1H-pyrazole-5-carbohydrazide
Catalogus # APOSPC410265-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :242.586 g/mol
Storage Temperature:Ambient
CAS Number:1001518-79-1
Formula:C₆H₆ClF₃N₄O
MDL Number:MFCD03419675