Order Entry
Netherlands
ContactUsLinkComponent
4-(4-Chlorophenyl)-2-(piperazin-1-yl)-6-(trifluoromethyl)pyrimidine dihydrochloride
4-(4-Chlorophenyl)-2-(piperazin-1-yl)-6-(trifluoromethyl)pyrimidine dihydrochloride
Catalog # APOSPC410122-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(4-Chlorophenyl)-2-(piperazin-1-yl)-6-(trifluoromethyl)pyrimidine dihydrochloride
Catalog # APOSPC410122-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :415.669 g/mol
Storage Temperature:Ambient
Formula:C₁₅H₁₄ClF₃N₄·2ClH
MDL Number:MFCD25371206