Order Entry
Netherlands
ContactUsLinkComponent
4-(3-Methoxyphenyl)-2-(piperazin-1-yl)-6-(trifluoromethyl)pyrimidine dihydrochloride
4-(3-Methoxyphenyl)-2-(piperazin-1-yl)-6-(trifluoromethyl)pyrimidine dihydrochloride
Catalog # APOSPC410119-1G
Supplier:  Apollo Scientific
CAS Number:  
4-(3-Methoxyphenyl)-2-(piperazin-1-yl)-6-(trifluoromethyl)pyrimidine dihydrochloride
Catalog # APOSPC410119-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :411.249 g/mol
Storage Temperature:Ambient
CAS Number:1431970-11-4
Formula:C₁₆H₁₇F₃N₄O·2ClH
MDL Number:MFCD25371203