Order Entry
Netherlands
ContactUsLinkComponent
3-Chloro-1-[5-(trifluoromethyl)pyridin-2-yl]-1H-pyrazol-4-amine hydrochloride
3-Chloro-1-[5-(trifluoromethyl)pyridin-2-yl]-1H-pyrazol-4-amine hydrochloride
Catalog # APOSPC410097-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Chloro-1-[5-(trifluoromethyl)pyridin-2-yl]-1H-pyrazol-4-amine hydrochloride
Catalog # APOSPC410097-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :299.08 g/mol
Storage Temperature:Ambient
Formula:C₉H₆ClF₃N₄·ClH
MDL Number:MFCD25371159