1-(Difluoromethyl)-1H-pyrazol-3-amine hydrochloride
Catalog # APOSPC410090-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:169.56 g/mol
Storage Temperature:Ambient
Formula:C₄H₅F₂N₃·ClH
MDL Number:MFCD25371147