Order Entry
Netherlands
ContactUsLinkComponent
 
1-(4-Fluorobenzyl)piperidine-3-carboxylic acid hydrochloride
Catalog # APOSPC410087-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(4-Fluorobenzyl)piperidine-3-carboxylic acid hydrochloride
Catalog # APOSPC410087-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :273.731 g/mol
Storage Temperature:Ambient
CAS Number:1185303-78-9
 :832737-45-8
Formula:C₁₃H₁₆FNO₂·ClH
MDL Number:MFCD06800730