1-[(2-Fluorophenoxy)methyl]-1H-pyrazol-4-amine hydrochloride
Catalog # APOSPC410085-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:243.665 g/mol
Storage Temperature:Ambient
CAS Number:1431963-66-4
Formula:C₁₀H₁₀FN₃O·ClH
MDL Number:MFCD25370980