4-Bromo-1-(4-fluoro-2-nitrophenyl)-1H-pyrazole
Catalog # APOSPC410069-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:286.057 g/mol
Storage Temperature:Ambient
Formula:C₉H₅BrFN₃O₂
MDL Number:MFCD09701699