Order Entry
Netherlands
ContactUsLinkComponent
3-Cyclopropyl-1-(4-fluorophenyl)-1H-pyrazol-5-amine hydrochloride
3-Cyclopropyl-1-(4-fluorophenyl)-1H-pyrazol-5-amine hydrochloride
Catalog # APOSPC410063-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Cyclopropyl-1-(4-fluorophenyl)-1H-pyrazol-5-amine hydrochloride
Catalog # APOSPC410063-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :253.703 g/mol
Storage Temperature:Ambient
Formula:C₁₂H₁₂FN₃·ClH
MDL Number:MFCD25371086