Order Entry
Netherlands
ContactUsLinkComponent
3-[1'-Ethyl-5-(trifluoromethyl)-1'H,2H-3,4'-bipyrazol-2-yl]propanenitrile
3-[1'-Ethyl-5-(trifluoromethyl)-1'H,2H-3,4'-bipyrazol-2-yl]propanenitrile
Catalog # APOSPC410044-1G
Supplier:  Apollo Scientific
CAS Number:  
3-[1'-Ethyl-5-(trifluoromethyl)-1'H,2H-3,4'-bipyrazol-2-yl]propanenitrile
Catalog # APOSPC410044-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :283.252 g/mol
Storage Temperature:Ambient
Formula:C₁₂H₁₂F₃N₅
MDL Number:MFCD09469286