Order Entry
Netherlands
ContactUsLinkComponent
1-[2-(4-Nitro-1H-pyrazol-1-yl)ethyl]-3-(trifluoromethyl)-1H-pyrazole
1-[2-(4-Nitro-1H-pyrazol-1-yl)ethyl]-3-(trifluoromethyl)-1H-pyrazole
Catalog # APOSPC410043-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[2-(4-Nitro-1H-pyrazol-1-yl)ethyl]-3-(trifluoromethyl)-1H-pyrazole
Catalog # APOSPC410043-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :275.187 g/mol
Storage Temperature:Ambient
Formula:C₉H₈F₃N₅O₂
MDL Number:MFCD09468206