Order Entry
Netherlands
ContactUsLinkComponent
1-(3-Fluorobenzyl)-1H-pyrazol-4-amine hydrochloride
1-(3-Fluorobenzyl)-1H-pyrazol-4-amine hydrochloride
Catalog # APOSPC410011-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(3-Fluorobenzyl)-1H-pyrazol-4-amine hydrochloride
Catalog # APOSPC410011-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :227.666 g/mol
Storage Temperature:Ambient
Formula:C₁₀H₁₀FN₃·ClH
MDL Number:MFCD04967265