4-Bromo-1-methyl-5-(trifluoromethyl)-1H-pyrazole-3-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:497833-03-1
:
Formula:C₆H₄BrF₃N₂O₂
MDL Number:MFCD03419817
:273.007 g/mol
Storage Temperature:Ambient
Frequently Bought Together