Order Entry
Netherlands
ContactUsLinkComponent
1-(4-Fluoro-benzyl)-2-piperidin-4-yl-1H-benzoimidazole hydrochloride
1-(4-Fluoro-benzyl)-2-piperidin-4-yl-1H-benzoimidazole hydrochloride
Catalog # APOSPC408416-500MG
Supplier:  Apollo Scientific
CAS Number:  
1-(4-Fluoro-benzyl)-2-piperidin-4-yl-1H-benzoimidazole hydrochloride
Catalog # APOSPC408416-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :345.842 g/mol
Storage Temperature:Ambient
CAS Number:1420975-03-6
Formula:C₁₉H₂₀FN₃·ClH
MDL Number:MFCD24369855