Piperidine-3-carboxylic acid cyclopropyl-(3-fluoro-benzyl)-amide hydrochloride
Catalog # APOSPC408410-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:312.81 g/mol
Storage Temperature:Ambient
CAS Number:1417793-24-8
Formula:C₁₆H₂₁FN₂O·ClH
MDL Number:MFCD22631640