Order Entry
Netherlands
ContactUsLinkComponent
3-(3-Chloro-5-trifluoromethyl-pyridin-2-yloxymethyl)-piperidine-1-carboxylic acid tert-butyl ester
3-(3-Chloro-5-trifluoromethyl-pyridin-2-yloxymethyl)-piperidine-1-carboxylic acid tert-butyl ester
Catalog # APOSPC408404-500MG
Supplier:  Apollo Scientific
CAS Number:  
3-(3-Chloro-5-trifluoromethyl-pyridin-2-yloxymethyl)-piperidine-1-carboxylic acid tert-butyl ester
Catalog # APOSPC408404-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :394.816 g/mol
Storage Temperature:Ambient
CAS Number:1417793-54-4
Formula:C₁₇H₂₂ClF₃N₂O₃
MDL Number:MFCD22631681