Order Entry
Netherlands
ContactUsLinkComponent
1-[1-(4-Fluoro-benzyl)-1H-benzoimidazol-2-yl]-piperidin-3-ol
1-[1-(4-Fluoro-benzyl)-1H-benzoimidazol-2-yl]-piperidin-3-ol
Catalog # APOSPC408384-500MG
Supplier:  Apollo Scientific
CAS Number:  
1-[1-(4-Fluoro-benzyl)-1H-benzoimidazol-2-yl]-piperidin-3-ol
Catalog # APOSPC408384-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :325.38 g/mol
Storage Temperature:Ambient
CAS Number:1417793-71-5
Formula:C₁₉H₂₀FN₃O
MDL Number:MFCD22631647