(R)-3-(4-Fluoro-benzenesulphonyl)-pyrrolidine-1-carboxylic acid tert-butyl ester
Catalog # APOSPC408344-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:329.387 g/mol
Storage Temperature:Ambient
CAS Number:1354010-97-1
Formula:C₁₅H₂₀FNO₄S
MDL Number:MFCD21098484