Order Entry
Netherlands
ContactUsLinkComponent
3-(4-Fluoro-phenylsulphanylmethyl)-piperidine hydrochloride
3-(4-Fluoro-phenylsulphanylmethyl)-piperidine hydrochloride
Catalog # APOSPC408332-500MG
Supplier:  Apollo Scientific
CAS Number:  
3-(4-Fluoro-phenylsulphanylmethyl)-piperidine hydrochloride
Catalog # APOSPC408332-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :261.786 g/mol
Storage Temperature:Ambient
CAS Number:1289385-78-9
Formula:C₁₂H₁₆FNS·ClH
MDL Number:MFCD18837558