Order Entry
Netherlands
ContactUsLinkComponent
(1-(5-Fluoropyrimidin-2-yl)piperidin-4-yl)methanol
(1-(5-Fluoropyrimidin-2-yl)piperidin-4-yl)methanol
Catalog # APOSPC408314-500MG
Supplier:  Apollo Scientific
CAS Number:  
(1-(5-Fluoropyrimidin-2-yl)piperidin-4-yl)methanol
Catalog # APOSPC408314-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :211.236 g/mol
Storage Temperature:Ambient
CAS Number:1032825-49-2
Formula:C₁₀H₁₄FN₃O
MDL Number:MFCD18380094