Order Entry
Netherlands
ContactUsLinkComponent
(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-piperidin-4-yl-amine hydrochloride
(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-piperidin-4-yl-amine hydrochloride
Catalog # APOSPC408302-500MG
Supplier:  Apollo Scientific
CAS Number:  
(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-piperidin-4-yl-amine hydrochloride
Catalog # APOSPC408302-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :316.15 g/mol
Storage Temperature:Ambient
CAS Number:1185315-66-5
Formula:C₁₁H₁₃ClF₃N₃·ClH
MDL Number:MFCD10698911