Methyl 2-fluoro-5-methoxybenzoate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:96826-42-5
:
Formula:C₉H₉FO₃
MDL Number:MFCD06203711
:184.164 g/mol
Boiling Pt:96…98 °C (2 mm Hg)
Storage Temperature:Ambient
Frequently Bought Together