Cyclopropyl-(5-fluoro-pyrimidin-2-yl)-amine
Catalog # APOSPC408018-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:153.157 g/mol
Storage Temperature:Ambient
CAS Number:1289387-32-1
Formula:C₇H₈FN₃
MDL Number:MFCD18837175