Order Entry
Netherlands
ContactUsLinkComponent
5,7-Dichloro-2-(trifluoromethyl)[1,2,4]triazolo[1,5-a]pyrimidine
5,7-Dichloro-2-(trifluoromethyl)[1,2,4]triazolo[1,5-a]pyrimidine
Catalog # APOSPC404016-250MG
Supplier:  Apollo Scientific
CAS Number:  
5,7-Dichloro-2-(trifluoromethyl)[1,2,4]triazolo[1,5-a]pyrimidine
Catalog # APOSPC404016-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :257 g/mol
Storage Temperature:Refrigerator
Formula:C₆HCl₂F₃N₄
MDL Number:MFCD32264096