(S)-2-(2,5-Difluorophenyl)pyrrolidine hydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1443624-23-4
:
Formula:C₁₀H₁₁F₂N·ClH
MDL Number:MFCD08751479
:219.659 g/mol
Storage Temperature:Ambient
Frequently Bought Together