
4-(2-Chloro-5-fluorophenyl)piperidine hydrochloride 95+%
Catalog # APOSPC402112-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:250.14 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1821329-21-8
Formula:C₁₁H₁₃ClFN·ClH
MDL Number:MFCD31629947