Order Entry
Netherlands
ContactUsLinkComponent
3-Fluoro-4-(piperidin-3-yl)pyridine dihydrochloride 97%
Irritant
3-Fluoro-4-(piperidin-3-yl)pyridine dihydrochloride 97%
Catalog # APOSPC402109-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-Fluoro-4-(piperidin-3-yl)pyridine dihydrochloride 97%
Catalog # APOSPC402109-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :253.144 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1260645-80-4
Formula:C₁₀H₁₃FN₂·2ClH
MDL Number:MFCD30099271