
(S)-2-(4-Fluorophenyl)piperidine hydrochloride 97%
Catalog # APOSPC402100-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:215.695 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1391431-07-4
Formula:C₁₁H₁₄FN·ClH
MDL Number:MFCD12910636