Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Fluoro-6-hydroxy-2,3-dihydro-1H-isoindol-1-one
5-Fluoro-6-hydroxy-2,3-dihydro-1H-isoindol-1-one
5-Fluoro-6-hydroxy-2,3-dihydro-1H-isoindol-1-one
APOSPC402049-250MG
:
Apollo Scientific
:
5-Fluoro-6-hydroxy-2,3-dihydro-1H-isoindol-1-one
APOSPC402049-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
167.137 g/mol
:
Ambient
:
1007455-25-5
:
C₈H₆FNO₂
:
MFCD12924344