Order Entry
Netherlands
ContactUsLinkComponent
7-(4-Fluorophenyl)-2,3-dihydro-1H-isoindol-1-one
7-(4-Fluorophenyl)-2,3-dihydro-1H-isoindol-1-one
Catalog # APOSPC402027-250MG
Supplier:  Apollo Scientific
CAS Number:  
7-(4-Fluorophenyl)-2,3-dihydro-1H-isoindol-1-one
Catalog # APOSPC402027-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :227.234 g/mol
Storage Temperature:Ambient
CAS Number:200049-49-6
Formula:C₁₄H₁₀FNO
MDL Number:MFCD13177841