3-Bromo-1-methyl-5-(trifluoromethyl)pyridin-2-one
Catalog # APOSPC401073-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:256.02 g/mol
Storage Temperature:Ambient
CAS Number:1215205-35-8
Formula:C₇H₅BrF₃NO
MDL Number:MFCD15143623