Methyl 2-bromo-2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]acetate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1672655-82-1
:
Formula:C₉H₆BrClF₃NO₂
MDL Number:MFCD28122852
:332.502 g/mol
Storage Temperature:Ambient
UN:1759
ADR:8,III
Frequently Bought Together