3-Cyano-2-hydroxy-6-methyl-4-(trifluoromethyl)pyridine
Catalog # APOSPC400624-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:202.133 g/mol
Storage Temperature:Ambient
Formula:C₈H₅F₃N₂O
MDL Number:MFCD00736353