Order Entry
Netherlands
ContactUsLinkComponent
4-Fluoropiperidine-4-carboxylic acid, N-BOC protected 98%
Irritant
4-Fluoropiperidine-4-carboxylic acid, N-BOC protected 98%
Catalog # APOSPC400525-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Fluoropiperidine-4-carboxylic acid, N-BOC protected 98%
Catalog # APOSPC400525-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :247.263 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:614731-04-3
Formula:C₁₁H₁₈FNO₄
MDL Number:MFCD08062530