Ethyl 3-{[(trifluoromethyl)sulphonyl]oxy}furo[2,3-c]pyridine-2-carboxylate
Catalog # APOSPC400501-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:339.245 g/mol
Storage Temperature:Ambient
CAS Number:927804-63-5
Formula:C₁₁H₈F₃NO₆S
MDL Number:MFCD22690758