
2-(Difluoromethoxy)pyridine-3-carbaldehyde
Catalog # APOSPC400160-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:173.117 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:2222104-51-8
Formula:C₇H₅F₂NO₂
MDL Number:MFCD31621142