
4-(Difluoromethoxy)pyridine-2-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1707365-38-5
:
Formula:C₇H₅F₂NO₃
MDL Number:MFCD27995942
:189.116 g/mol
:GHS07
Storage Temperature:Kamertemperatuur
Frequently Bought Together