
Methyl 2-[1-methyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]benzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:898289-62-8
:
Formula:C₁₃H₁₁F₃N₂O₂
MDL Number:MFCD09879929
:284.234 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together