2-Benzyl-4-chloro-5-[(trifluoromethyl)sulphonyloxy]-2H-pyridazin-3-one
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:856250-49-2
:
Formula:C₁₂H₈ClF₃N₂O₄S
MDL Number:MFCD07367011
:368.716 g/mol
Storage Temperature:Ambient
Frequently Bought Together