Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-(Trifluoromethyl)pyridine-2-carboxamide
6-(Trifluoromethyl)pyridine-2-carboxamide
6-(Trifluoromethyl)pyridine-2-carboxamide
APOSPC32880-1G
:
Apollo Scientific
:
6-(Trifluoromethyl)pyridine-2-carboxamide
APOSPC32880-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
190.123 g/mol
:
Ambient
:
22245-84-7
:
C₇H₅F₃N₂O
:
MFCD02180820