
1-(Fluorosulphonyl)-2,3-dimethyl-1H-imidazol-3-ium trifluoromethanesulphonate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2179072-33-2
:
Formula:C₅H₈FN₂O₂S·CF₃O₃S
MDL Number:MFCD31693095
:328.262 g/mol
Melting Pt:60…62 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together