
1,1,1,3,3,3-Hexafluoro-2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)propan-2-ol 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:449804-09-5
:
Formula:C₁₅H₁₇BF₆O₃
MDL Number:MFCD22570625
:370.095 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together