Order Entry
Netherlands
ContactUsLinkComponent
2,6-Dichloro-5-fluoro-N-pyridin-3-ylnicotinamide
Irritant
2,6-Dichloro-5-fluoro-N-pyridin-3-ylnicotinamide
Catalog # APOSPC32692-100MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,6-Dichloro-5-fluoro-N-pyridin-3-ylnicotinamide
Catalog # APOSPC32692-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :286.089 g/mol
Melting Pt:133…134 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:680217-89-4
Formula:C₁₁H₆Cl₂FN₃O
MDL Number:MFCD01934909