
2,6-Dichloro-5-fluoro-N-(thien-2-ylmethyl)nicotinamide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:680217-82-7
:
Formula:C₁₁H₇Cl₂FN₂OS
MDL Number:MFCD01765455
:305.155 g/mol
Melting Pt:145…146 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together