Order Entry
Netherlands
ContactUsLinkComponent
4-(Bromomethyl)-1-methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazole
Corrosive
Irritant
4-(Bromomethyl)-1-methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazole
Catalog # APOSPC3229-1G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
4-(Bromomethyl)-1-methyl-5-phenoxy-3-(trifluoromethyl)-1H-pyrazole
Catalog # APOSPC3229-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :335.12 g/mol
Melting Pt:75.5…77.5 °C
Storage Temperature:Ambient
UN:3261
ADR:8,III
 :GHS05
 :GHS07
CAS Number:912569-72-3
Formula:C₁₂H₁₀BrF₃N₂O
MDL Number:MFCD09879947